missing translation for 'onlineSavingsMsg'
Learn More
tert-Butyl methyl ether, 99+%, Extra Dry over Molecular Sieve, AcroSeal™, Thermo Scientific Chemicals Product Code.: 10628273

tert-Butyl methyl ether, 99+%, Extra Dry over Molecular Sieve, AcroSeal™, Thermo Scientific Chemicals

Product Code. 10628273
100 mL, AcroSeal™ Glass Bottle
Click to view available options
Quantity:
100 mL
1 L
Packaging:
AcroSeal™ Glass Bottle
This item is not returnable. View return policy
This item is packed in AcroSeal™, the industry-leading packaging for air and moisture sensitive reagents.
Find out more about AcroSeal™ packaging.
 

Product Code. 10628273

Brand: Thermo Scientific Acros 375221000

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
This item is packed in AcroSeal™, the industry-leading packaging for air and moisture sensitive reagents.
Find out more about AcroSeal™ packaging.
 

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

Molecular Formula C5H12O
Molecular Weight (g/mol) 88.15
MDL Number MFCD00008812
InChI Key BZLVMXJERCGZMT-UHFFFAOYSA-N
Synonym tert-butyl methyl ether, methyl tert-butyl ether, mtbe, methyl t-butyl ether, t-butyl methyl ether, methyl tertiary-butyl ether, propane, 2-methoxy-2-methyl, methyl-tert-butyl ether, methyl-t-butyl ether, 2-methyl-2-methoxypropaneShow MoreShow Less
PubChem CID 15413
ChEBI CHEBI:27642
IUPAC Name 2-methoxy-2-methylpropane
SMILES CC(C)(C)OC

Specifications

Melting Point -110.0°C
CAS Min % 99
Color Colorless
CAS Max % 100
Density 0.7404g/mL
Boiling Point 54.0°C to 56.0°C
Quantity 100 mL
Flash Point −28°C
Infrared Spectrum Authentic
Packaging AcroSeal™ Glass Bottle
Physical Form Liquid
Refractive Index 1.3675 to 1.3695
Assay Percent Range 99% min. (GC)
Specific Gravity 0.7404
Linear Formula (CH3)3COCH3
Viscosity 0.36 mPa.s (20°C)
Water 0.005% max.
Merck Index 15, 6106
Solubility Information Solubility in water: 51g/L (20°C). Other solubilities: miscible with most organic solvents, unstable in acid solutions, soly of water in methyl-t-butyl ether : 1,5g/100g
Formula Weight 88.15
Percent Purity 99+%
Grade Extra Dry over Molecular Sieve
Chemical Name or Material tert-Butyl methyl ether, Over Molecular Sieves
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number2496107 Certificate TypeCertificate of Analysis Date03/02/2025 Catalog Number375220010,375221000
Lot Number2496106 Certificate TypeCertificate of Analysis Date31/01/2025 Catalog Number375220010,375221000
Lot Number2405744 Certificate TypeCertificate of Analysis Date31/01/2025 Catalog Number375220010,375221000
Lot Number2416303 Certificate TypeCertificate of Analysis Date30/01/2025 Catalog Number375220010,375221000
Lot Number2416302 Certificate TypeCertificate of Analysis Date30/01/2025 Catalog Number375220010,375221000
compliance-icons compliance-icons
Product Identifier
  • tert-Butyl methyl ether
Signal Word
  • Danger
Hazard Category
  • Flammable liquid Category 2
  • Skin corrosion/irritation Category 2
Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H315-Causes skin irritation.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P240-Ground/bond container and receiving equipment.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
tert-Butyl methyl ether, 99+%, Extra Dry over Molecular Sieve, AcroSeal™, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.