missing translation for 'onlineSavingsMsg'
Learn More
Learn More
tert-Butyl N-(2-cyanoethyl)carbamate, 97%, Thermo Scientific™
Click to view available options
Quantity:
1 g
10 g
Packaging:
Amber Glass Bottle
Specifications
Specifications
CAS Min % | 97.0 |
Assay Percent Range | 97% |
Linear Formula | (CH3)3COC(O)NH(CH2)2CN |
Packaging | Amber Glass Bottle |
Formula Weight | 170.21 |
Percent Purity | 97% |
Quantity | 10 g |
Chemical Name or Material | Tert-butyl N-(2-cyanoethyl)carbamate |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
tert-Butyl N-(2-cyanoethyl)carbamate, 97%, Thermo Scientific™ >
Spot an opportunity for improvement?Share a Content Correction