missing translation for 'onlineSavingsMsg'
Learn More
tert-Butyl N-[4-(aminomethyl)phenyl]carbamate, 97%, Thermo Scientific™ Product Code.: 10390773

tert-Butyl N-[4-(aminomethyl)phenyl]carbamate, 97%, Thermo Scientific™

Product Code. 10390773
1 g
Click to view available options
Quantity:
250 mg
1 g
10 g
This item is not returnable. View return policy

Product Code. 10390773

Brand: Thermo Scientific Maybridge MO01028DA

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 220298-96-4
Molecular Formula C12H18N2O2
Molecular Weight (g/mol) 222.29
MDL Number MFCD02183573
InChI Key URXUHALBOWYXJZ-UHFFFAOYSA-N
Synonym tert-butyl n-4-aminomethyl phenyl carbamate, 4-aminomethyl-1-n-boc-aniline, tert-butyl 4-aminomethyl phenyl carbamate, 4-boc-amino benzylamine, 4-aminomethy-1-n-boc-aniline, 4-n-boc-amino benzylamine, 4-tert-butoxycarbonylamino benzylamine, 4-aminomethyl-n-boc-aniline, tert-butyl-n-4-aminomethyl phenyl carbamateShow MoreShow Less
PubChem CID 2794659
IUPAC Name tert-butyl N-[4-(aminomethyl)phenyl]carbamate
SMILES CC(C)(C)OC(=O)NC1=CC=C(CN)C=C1

Specifications

CAS Min % 97.0
Assay Percent Range 95%
Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 222.29
Percent Purity 97%
Chemical Name or Material tert-Butyl N-[4-(aminomethyl)phenyl]carbamate
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
tert-Butyl N-[4-(aminomethyl)phenyl]carbamate, 97%, Thermo Scientific™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.