missing translation for 'onlineSavingsMsg'
Learn More
Learn More
tert-Butyl N-(4-piperidinylmethyl)carbamate, 97%, Thermo Scientific™
Click to view available options
Quantity:
250 mg
1 g
25 g
Specifications
Specifications
Assay Percent Range | 95% |
Quantity | 250 mg |
Formula Weight | 214.31 |
Percent Purity | 95% |
Chemical Name or Material | tert-Butyl N-(4-piperidinylmethyl)carbamate |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
tert-Butyl N-(4-piperidinylmethyl)carbamate, 97%, Thermo Scientific™ >
Spot an opportunity for improvement?Share a Content Correction