missing translation for 'onlineSavingsMsg'
Learn More

tert-Butylamine, 98%, Thermo Scientific Chemicals

36.78 € - 315.57 €

Chemical Identifiers

CAS 75-64-9
Molecular Formula C4H11N
MDL Number MFCD00008050
InChI Key YBRBMKDOPFTVDT-UHFFFAOYSA-N
Synonym tert-butylamine, erbumine, 2-amino-2-methylpropane, t-butylamine, 2-propanamine, 2-methyl, trimethylaminomethane, 1,1-dimethylethylamine, 2-aminoisobutane, 1,1-dimethylethanamine, butylamine, tert
PubChem CID 6385
ChEBI CHEBI:44639
IUPAC Name 2-methylpropan-2-amine
SMILES CC(C)(C)N
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11469033
View Documents
Thermo Scientific Alfa Aesar
A11135.AP
500 mL
36.78 €
500mL
Please to purchase this item. Need a web account? Register with us today!
11479033
View Documents
Thermo Scientific Alfa Aesar
A11135.0F
2500 mL
114.57 €
2500mL
Please to purchase this item. Need a web account? Register with us today!
11489033
View Documents
Thermo Scientific Alfa Aesar
A11135.0D
10,000 mL
315.57 €
10000mL
Please to purchase this item. Need a web account? Register with us today!
Description

Description

tert-butylamine is used as a precursor for the preparation of rubber accelerators, pharmaceuticals and dyes. It is employed in synthesis of N-tert-butyl-acetamide, isoquinolines, pyridines, decumbenine, sulfontes, sulfonamides and methyl beta-ketophosphonate. It is also used in the study the effect of amines in influencing atmospheric sulfuric acid - water nucleation, hydroamination of terminal alkynes and measuring the surface acidity of solid catalyst. It acts as an indicator molecule in the study of NMR -based pH determination method.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
tert-butylamine is used as a precursor for the preparation of rubber accelerators, pharmaceuticals and dyes. It is employed in synthesis of N-tert-butyl-acetamide, isoquinolines, pyridines, decumbenine, sulfontes, sulfonamides and methyl beta-ketophosphonate. It is also used in the study the effect of amines in influencing atmospheric sulfuric acid - water nucleation, hydroamination of terminal alkynes and measuring the surface acidity of solid catalyst. It acts as an indicator molecule in the study of NMR -based pH determination method.

Solubility
Miscible with water, alcohol, ether, chloroform and organic solvents.

Notes
Air sensitive. Incompatible with strong oxidizing agents, strong acids and acid anhydrides.
Specifications

Chemical Identifiers

75-64-9
MFCD00008050
tert-butylamine, erbumine, 2-amino-2-methylpropane, t-butylamine, 2-propanamine, 2-methyl, trimethylaminomethane, 1,1-dimethylethylamine, 2-aminoisobutane, 1,1-dimethylethanamine, butylamine, tert
CHEBI:44639
CC(C)(C)N
C4H11N
YBRBMKDOPFTVDT-UHFFFAOYSA-N
6385
2-methylpropan-2-amine

Specifications

75-64-9
0.696
−38°C (−36°F)
C4H11N
MFCD00008050
605267
14,1545
Miscible with water,alcohol,ether,chloroform and organic solvents.
CC(C)(C)N
73.139
CHEBI:44639
98%
-67°C
44°C to 46°C
Amine-like
1.378
UN2733
Air Sensitive
tert-butylamine, erbumine, 2-amino-2-methylpropane, t-butylamine, 2-propanamine, 2-methyl, trimethylaminomethane, 1,1-dimethylethylamine, 2-aminoisobutane, 1,1-dimethylethanamine, butylamine, tert
YBRBMKDOPFTVDT-UHFFFAOYSA-N
2-methylpropan-2-amine
6385
73.14
tert-Butylamine
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H225-H300-H314-H318-H332
Highly flammable liquid and vapor.
Fatal if swallowed.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if inhaled.

P210-P233-P235-P240-P241-P242-P243-P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P370+P378q-P501c

H225-H302-H314-H331-H335-H373

missing translation for 'dotInformation' : Hazard Class: 8; Packaging Group: I

missing translation for 'einecsNumber' : 200-888-1

missing translation for 'rtecsNumber' : EO3330000

missing translation for 'tsca' : Yes

Recommended Storage : Ambient temperatures

Product Identifier
  • tert-Butylamine

Signal Word
  • Danger

Hazard Category
  • Acute toxicity Category 3
  • Acute toxicity Category 4
  • LONG-TERM AQUATIC HAZARD Chronic 3
  • Serious eye damage/eye irritation Category 1
  • Flammable liquid Category 2
  • Skin corrosion/irritation Category 1 A

Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H302-Harmful if swallowed.
  • H314-Causes severe skin burns and eye damage.
  • H318-Causes serious eye damage.
  • H331-Toxic if inhaled.
  • H412-Harmful to aquatic life with long lasting effects.

Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

Supplemental information
  • MIXTURE LIST-Contains : tert-Butylamine

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.