Learn More
Terumo™ Hypodermic Needles
Provides ultra sharp needles, which translate into greater comfort for the patient.
Brand:  Terumo™ TUAN2125R1
 View more versions of this product
Product Code. 12810126
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Explore more special offers
Description
Description
Terumo™ Hypodermic Needles
Specifications
Specifications
21 G
1 in.
Ultra-thin Wall
Yes
Double Bevel Design