Learn More
Tetrabutylammonium fluoride, 1M solution in THF, containing ca. 5% water, Thermo Scientific Chemicals
Brand: Thermo Scientific 201950050
30.40 EUR valid until 2025-12-31
Use promo code "25339" to get your promotional price.
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
C-X bond formation reagent
Specifications
Tetrabutylammonium fluoride | |
429-41-4, 109-99-9, 7732-18-5 | |
[CH3(CH2)3]4NF | |
04,477; 05,645; 07,353; 08,467; 09,444; 10,378; 11,499; 12,458; 14,293; 15,168; 17,363 | |
tetrabutylammonium fluoride, tbaf, tetrabutylazanium fluoride, tetrabutyl ammonium fluoride, tetra-n-butylammonium fluoride, tetrabutylamine, fluoride, n,n,n-tributylbutan-1-aminium fluoride, 1-butanaminium, n,n,n-tributyl-, fluoride, n,n,n-tributyl-1-butanaminium fluoride, 1-butanaminium, n,n,n-tributyl-, fluoride 1:1 | |
[F-].CCCC[N+](CCCC)(CCCC)CCCC | |
261.47 | |
CHEBI:51990 | |
Glass bottle | |
5 mL |
1M Solution in THF, Containing ca 5% Water | |
C16H36FN | |
MFCD00011747 | |
0.8870g/mL | |
FPGGTKZVZWFYPV-UHFFFAOYSA-M | |
tetrabutylazanium;fluoride | |
2724141 | |
261.46 | |
0.887 | |
−17°C |
Safety and Handling
- Tetrabutylammonium fluoride, 1M solution in THF, containing ca. 5% water
Signal Word
- Danger
Hazard Category
- Carcinogenicity Category 2
- Serious eye damage/eye irritation Category 1
- Flammable liquid Category 2
- Skin corrosion/irritation Category 1 B
- Specific target organ toxicity Category 3
Hazard Statement
- H318-Causes serious eye damage.
- H314-Causes severe skin burns and eye damage.
- H351-Suspected of causing cancer.
- H335-May cause respiratory irritation.
- H336-May cause drowsiness or dizziness.
- H225-Highly flammable liquid and vapour.
- EUH019-May form explosive peroxides.
Precautionary Statement
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
AB4620021 | Certificate of Analysis | 27/02/2025 | |
A0454699 | Certificate of Analysis | 31/01/2025 | 201951000,201955000,201950050,201950025 |
AA4534301 | Certificate of Analysis | 31/01/2025 | |
A0453755 | Certificate of Analysis | 03/01/2024 | 201951000,201955000,201950050,201950025 |
A0447776 | Certificate of Analysis | 14/12/2023 | 201951000,201955000,201950050,201950025 |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
RUO – Research Use Only