Learn More
Tetrabutylammonium fluoride, 1M solution in THF, containing ca. 5% water, Thermo Scientific Chemicals
29.20 € - 1454.00 €
Specifications
Chemical Name or Material | Tetrabutylammonium fluoride |
---|---|
Name Note | 1M Solution in THF, Containing ca 5% Water |
CAS | 429-41-4, 109-99-9, 7732-18-5 |
Molecular Formula | C16H36FN |
Linear Formula | [CH3(CH2)3]4NF |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10378480
|
Thermo Scientific
201950050 |
5 mL | Glass bottle |
29.20 €
5mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10676402
|
Thermo Scientific Acros
201951000 |
100 mL | Glass bottle |
122.00 €
100mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10686402
|
Thermo Scientific Acros
201955000 |
500 mL | Glass bottle |
420.00 €
500mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
11905451
|
Thermo Scientific Acros
201950025 |
2.5 L | Glass bottle |
1454.00 €
2.50 L |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
C-X bond formation reagentSpecifications
Tetrabutylammonium fluoride | |
429-41-4, 109-99-9, 7732-18-5 | |
[CH3(CH2)3]4NF | |
04,477; 05,645; 07,353; 08,467; 09,444; 10,378; 11,499; 12,458; 14,293; 15,168; 17,363 | |
tetrabutylammonium fluoride, tbaf, tetrabutylazanium fluoride, tetrabutyl ammonium fluoride, tetra-n-butylammonium fluoride, tetrabutylamine, fluoride, n,n,n-tributylbutan-1-aminium fluoride, 1-butanaminium, n,n,n-tributyl-, fluoride, n,n,n-tributyl-1-butanaminium fluoride, 1-butanaminium, n,n,n-tributyl-, fluoride 1:1 | |
[F-].CCCC[N+](CCCC)(CCCC)CCCC | |
261.47 | |
CHEBI:51990 | |
Glass bottle | |
−17°C |
1M Solution in THF, Containing ca 5% Water | |
C16H36FN | |
MFCD00011747 | |
0.8870g/mL | |
FPGGTKZVZWFYPV-UHFFFAOYSA-M | |
tetrabutylazanium;fluoride | |
2724141 | |
261.46 | |
0.887 |
Safety and Handling
GHS H Statement
Causes severe skin burns and eye damage. Suspected of causing cancer. May cause respiratory irritation. Highly flammable liquid and vapor. May form explosive peroxides. May cause drowsiness or dizziness.
GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces. - No smoking. IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/shower. Wear protective gloves/protective clothing/eye protection/face protection. IF SWALLOWED: rinse mouth. Do NOT induce vomiting. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. Immediately call a POISON CENTER or doctor/physician.
Danger
missing translation for 'einecsNumber' : 207-057-2
missing translation for 'tsca' : TSCA
- Tetrabutylammonium fluoride, 1M solution in THF, containing ca. 5% water
Signal Word
- Danger
Hazard Category
- Carcinogenicity Category 2
- Serious eye damage/eye irritation Category 1
- Flammable liquid Category 2
- Skin corrosion/irritation Category 1 B
- Specific target organ toxicity Category 3
Hazard Statement
- H318-Causes serious eye damage.
- H314-Causes severe skin burns and eye damage.
- H351-Suspected of causing cancer.
- H335-May cause respiratory irritation.
- H336-May cause drowsiness or dizziness.
- H225-Highly flammable liquid and vapour.
- EUH019-May form explosive peroxides.
Precautionary Statement
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
RUO – Research Use Only