missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tetrachloroethylene, Honeywell
85.50 € - 6596.26 €
Chemical Identifiers
CAS | 127-18-4 |
---|---|
Molecular Formula | C2Cl4 |
Molecular Weight (g/mol) | 165.82 |
MDL Number | MFCD00000834 |
InChI Key | CYTYCFOTNPOANT-UHFFFAOYSA-N |
Synonym | tetrachloroethylene, tetrachloroethene, perchloroethylene, perchlorethylene, perc, ethene, tetrachloro, tetrachlorethylene, ethylene tetrachloride, carbon dichloride, ankilostin |
PubChem CID | 31373 |
ChEBI | CHEBI:17300 |
SMILES | ClC(Cl)=C(Cl)Cl |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15621550
|
Honeywell Research Chemicals
16211-1L |
1 L | N/A | ||||||
15631550
|
Honeywell Research Chemicals
16211-2.5L |
2.5 L | N/A | ||||||
15611550
|
Honeywell Research Chemicals
16211-10L |
10 L | N/A | ||||||
15661550
|
Honeywell Research Chemicals
16211-800KG |
800 kg | N/A | ||||||
Descripción
Identificadores de productos químicos
127-18-4 | |
165.82 | |
CYTYCFOTNPOANT-UHFFFAOYSA-N | |
31373 | |
ClC(Cl)=C(Cl)Cl |
C2Cl4 | |
MFCD00000834 | |
tetrachloroethylene, tetrachloroethene, perchloroethylene, perchlorethylene, perc, ethene, tetrachloro, tetrachlorethylene, ethylene tetrachloride, carbon dichloride, ankilostin | |
CHEBI:17300 |
Especificaciones
127-18-4 | |
C2Cl4 | |
MFCD00000834 | |
1361721 | |
CYTYCFOTNPOANT-UHFFFAOYSA-N | |
1,1,2,2-tetrachloroethene | |
31373 | |
CHEBI:17300 | |
13mmHg (20°C), 19mmHg (25°C) | |
Tetrachloroethylene |
1.623g/mL at 25°C | |
CCl2=CCl2 | |
1897 | |
tetrachloroethylene, tetrachloroethene, perchloroethylene, perchlorethylene, perc, ethene, tetrachloro, tetrachlorethylene, ethylene tetrachloride, carbon dichloride, ankilostin | |
ClC(Cl)=C(Cl)Cl | |
165.82 | |
5.83 (vs air) | |
165.83g/mol | |
≥99.5% |
Seguridad y manipulación
P280-P305 + P351 + P338
H315-H317-H319-H336-H351-H411
Warning
missing translation for 'einecsNumber' : 204-825-9
missing translation for 'rtecsNumber' : KX3850000
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto