missing translation for 'onlineSavingsMsg'
Learn More

WTW™ TetraCon™ 325 Standard Conductivity Cell

Use these conductivity cells for precise and reliable measurements in any conditions. WTW™ TetraCon™ 325 Standard Conductivity Cells feature 4 electrode technology makes them insensitive to pollution, uninfluenced by polarization effects.

Brand:  WTW™ 301960

580.00 EUR valid until 2025-12-31
Use promo code "25339" to get your promotional price.


Product Code. 10069160

  • 624.00 € / Each
In Stock
to see stock.
Edge
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • 4 electrode graphite cell with 1.5 m cable
  • No measuring errors due to cable, polarization effects or changes of the electrical edge field
  • Highest precision and linearity
  • Long-term stable cell constant thanks to graphite electrodes
TRUSTED_SUSTAINABILITY
Specifications

Specifications

0.475 cm-1
Standard Conductivity Cell
1.5 m
4-electrode, temperature sensor, 1.5m Length cable and waterproof plug
TetraCon™ 325
0°C to 90°C
15.3 mm
Epoxy, Graphite
IP68
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
WTW™ TetraCon™ 325 Standard Conductivity Cell > Material: Epoxy/Graphite; Measuring Range: 1mu S/cm to 1S/cm; Temperature Range: 0 to 90C; Cell Constant: 0.475cm

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.