missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tetrahydrofuran, for HPLC, Unstabilised, Fisher Chemical™
C4H8O, CAS Number-109-99-9, butylene oxide, tetraidrofurano, 1,4-epoxybutane, furanidine, oxacyclopentane, tetrahydrofuranne, hydrofuran, furan, tetrahydro, tetrahydrofuran, tetramethylene oxide, 2.5L, 66 deg.C, CHEBI:26911, Colorless, 72.11g/mol, WYURNTSHIVDZCO-UHFFFAOYSA-N, oxolane, -108.4 deg.C
465.00 € - 575.00 €
Chemical Identifiers
CAS | 109-99-9 |
---|---|
Molecular Formula | C4H8O |
Molecular Weight (g/mol) | 72.11 |
MDL Number | MFCD00005356 |
InChI Key | WYURNTSHIVDZCO-UHFFFAOYSA-N |
Synonym | tetrahydrofuran, furan, tetrahydro, butylene oxide, furanidine, hydrofuran, tetramethylene oxide, oxacyclopentane, 1,4-epoxybutane, tetrahydrofuranne, tetraidrofurano |
PubChem CID | 8028 |
ChEBI | CHEBI:26911 |
IUPAC Name | oxolane |
SMILES | C1CCOC1 |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10264350
|
Fisher Chemical
T/0706/PB15 |
1 L | Plastic-coated Glass Bottle |
465.00 €
1 L |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10578070
|
Fisher Chemical
T/0706/PB17 |
2.5 L | Plastic-coated Glass Bottle |
575.00 €
2.50 L |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
C4H8O, CAS Number-109-99-9, butylene oxide, tetraidrofurano, 1,4-epoxybutane, furanidine, oxacyclopentane, tetrahydrofuranne, hydrofuran, furan, tetrahydro, tetrahydrofuran, tetramethylene oxide, 2.5L, 66 deg.C, CHEBI:26911, Colorless, 72.11g/mol, WYURNTSHIVDZCO-UHFFFAOYSA-N, oxolane, -108.4 deg.C
Chemical Identifiers
109-99-9 | |
72.11 | |
WYURNTSHIVDZCO-UHFFFAOYSA-N | |
8028 | |
oxolane |
C4H8O | |
MFCD00005356 | |
tetrahydrofuran, furan, tetrahydro, butylene oxide, furanidine, hydrofuran, tetramethylene oxide, oxacyclopentane, 1,4-epoxybutane, tetrahydrofuranne, tetraidrofurano | |
CHEBI:26911 | |
C1CCOC1 |
Specifications
7 to 8 | |
109-99-9 | |
66°C | |
C4H8O | |
200mbar at 20°C | |
tetrahydrofuran, furan, tetrahydro, butylene oxide, furanidine, hydrofuran, tetramethylene oxide, oxacyclopentane, 1,4-epoxybutane, tetrahydrofuranne, tetraidrofurano | |
C1CCOC1 | |
72.11 | |
CHEBI:26911 |
-108.4°C | |
Colorless | |
Liquid | |
MFCD00005356 | |
0.55cp at 20°C | |
WYURNTSHIVDZCO-UHFFFAOYSA-N | |
oxolane | |
8028 | |
72.11g/mol |
Safety and Handling
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
- Tetrahydrofuran
Signal Word
- Danger
Hazard Category
- Acute toxicity Category 4
- Carcinogenicity Category 2
- Serious eye damage/eye irritation Category 2
- Flammable liquid Category 2
- Specific target organ toxicity Category 3
Hazard Statement
- H319-Causes serious eye irritation.
- H351-Suspected of causing cancer.
- H335-May cause respiratory irritation.
- H336-May cause drowsiness or dizziness.
- H302-Harmful if swallowed.
- H225-Highly flammable liquid and vapour.
- EUH019-May form explosive peroxides.
Precautionary Statement
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P312-Call a POISON CENTER/doctor/A§/if you feel unwell.
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title