Learn More
Molecular Probes™ Tetramethylrhodamine-5-(and-6)-Isothiocyanate (5(6)-TRITC), mixed isomers
Tetramethylrhodamine-5-(and-6)-Isothiocyanate (5(6)-TRITC), mixed isomers
Brand: Molecular Probes™ T490
285.00 EUR valid until 2025-03-28
Use promo code "24107" to get your promotional price.
356.25 EUR valid until 2025-05-31
Use promo code "25977" to get your promotional price.
Description
The thiol-reactive tetramethylrhodamine-5-(and-6)-isothiocyanate (5(6)-TRITC) can be used to can be used to create bright orange-red-fluorescent bioconjugates with excitation/emission maxima ∼555/580.
Specifications
10 mg | |
Store in freezer (-5 to -30°C) and protect from light. | |
Room Temperature | |
TMR (Tetramethylrhodamine) | |
580 |
Tetramethylrhodamine-5-(and-6)-Isothiocyanate | |
Amine | |
Isothiocyanate | |
Classic Dyes | |
555 |
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only. Not for use in diagnostic procedures.