missing translation for 'onlineSavingsMsg'
Learn More

Texwipe™ Vertex™ Wipers

For maximum abrasion resistance.

Brand:  Texwipe™ TX42P

333.95 EUR valid until 2025-02-13
Use promo code "24130" to get your promotional price.



 View more versions of this product

Product Code. 17989710

  • 449.00 € / Pack of 200

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

 

  • Made of 100% polyester
  • Pre-wetted wiper
  • Double-knit design
  • Have sealed laser edge
  • Nonsterile
  • Available in High Durability, High Sorption and the Microdenier Series
  • Sorbency capacity is between 350 and 450mL/m2

 

Specifications

Specifications

Pre-wetted Wiper
30.5 cm
30.5 cm
ISO 9001:2000, 13485:2003, 14001:2004 and OHSAS 18001:1999
Sharp sealed laser
Ideal for use on surfaces with sharp edges and spill control
70% Isopropyl Alcohol, 30% Deionised Water
12 in.
12 in.
Polyester
Double-knit
12 x 12 in. (30.48 x 30.48 cm)
50/Bag
Non-sterile
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

Product Identifier
  • Texwipe Wipers pre-wetted with 60% - 70 % IPA

Signal Word
  • Warning

Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Flammable solid Category 2
  • Specific target organ toxicity Category 3

Hazard Statement
  • H228-Flammable solid.
  • H319-Causes serious eye irritation.
  • H336-May cause drowsiness or dizziness.

Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P264-Wash hands thoroughly after handling.
  • P271-Use only outdoors or in a well-ventilated area.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
  • P405-Store locked up.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.

Supplemental information
  • MIXTURE LIST-Contains : propan-2-ol

SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.