missing translation for 'onlineSavingsMsg'
Learn More

Thallium(I) hexafluorophosphate(V), 97% min, Thermo Scientific Chemicals

269.34 € - 1035.15 €

Chemical Identifiers

CAS 60969-19-9
Molecular Formula F6PTl
Molecular Weight (g/mol) 349.344
MDL Number MFCD00049807
InChI Key FRZBCOUMLHRKRT-UHFFFAOYSA-N
Synonym thallium hexafluorophosphate, tlpf6, thallium i hexafluorophosphate, thallium 1+ hexafluorophosphate, thallium 1+ hexafluoro-$l^ 5-phosphanuide, $l^ 1-thallanylium hexafluoro-$l^ 5-phosphanuide
PubChem CID 10904204
IUPAC Name thallium(1+);hexafluorophosphate
SMILES F[P-](F)(F)(F)(F)F.[Tl+]
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11354577
SDS View Documents
Thermo Scientific Alfa Aesar
039627.06
5 g
269.34 €
5g
Please to purchase this item. Need a web account? Register with us today!
11344577
SDS View Documents
Thermo Scientific Alfa Aesar
039627.14
25 g
1035.15 €
25g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications
Show More
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H300-H330-H373
Fatal if swallowed.
Fatal if inhaled.
May cause damage to organs through prolonged or repeated exposure.

P260-P264b-P270-P271-P284-P301+P310-P304+P340-P310-P314-P330-P501c

H300+H330-H373

missing translation for 'dotInformation' : Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: THALLIUM COMPOUNDS, N.O.S.

missing translation for 'tsca' : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Thallium(I) hexafluorophosphate(V), 97% min, Thermo Scientific Chemicals

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.