missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thallous Acetate, 99%, Thermo Scientific Chemicals
Click to view available options
Quantity:
25 g
100 g
500 g
Packaging:
Glass bottle
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications
Specifications
Melting Point | 128.0°C to 130.0°C |
Color | White |
Physical Form | Fine Crystalline Powder |
Quantity | 100 g |
Assay Percent Range | 99% |
Linear Formula | CH3COOTl |
Merck Index | 15, 9405 |
Formula Weight | 263.42 |
Percent Purity | 99% |
Infrared Spectrum | Authentic |
Show More |
Product Suggestions
Customers who viewed this item also viewed
Viewing 1-4 of
Certificates
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
5 results found
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
A0468042 | Certificate of Analysis | 31/01/2025 | 420771000,420775000,420770250 |
A0465547 | Certificate of Analysis | 20/08/2024 | 420771000,420770250,420775000 |
A0459650 | Certificate of Analysis | 25/01/2024 | 420771000,420770250,420775000 |
A0401256 | Certificate of Analysis | 14/12/2022 | 420771000,420770250,420775000 |
A0398223 | Certificate of Analysis | 14/12/2022 | 420771000,420770250,420775000 |
Safety and Handling



Product Identifier
- Thallous acetate
- Danger
- Acute toxicity Category 1
- Acute toxicity Category 2
- LONG-TERM AQUATIC HAZARD Chronic 2
- Specific target organ toxicity after repeated exposure Category 2
- H310-Fatal in contact with skin.
- H330-Fatal if inhaled.
- H373-May cause damage to organs.
- H411-Toxic to aquatic life with long lasting effects.
- H300-Fatal if swallowed.
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P273-Avoid release to the environment.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P310-Immediately call a POISON CENTER/doctor/.
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Thallous Acetate, 99%, Thermo Scientific Chemicals >
Spot an opportunity for improvement?Share a Content Correction