missing translation for 'onlineSavingsMsg'
Learn More
Thallous Acetate, 99%, Thermo Scientific Chemicals Product Code.: 10577942

Thallous Acetate, 99%, Thermo Scientific Chemicals

Product Code. 10577942
25 g, Glass bottle
Click to view available options
Quantity:
25 g
100 g
500 g
Packaging:
Glass bottle
This item is not returnable. View return policy

Product Code. 10577942

Brand: Thermo Scientific Acros 420770250

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

Currently Unavailable: We have no information when this product will be available
View alternative products
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 563-68-8
Molecular Formula C2H3O2Tl
Molecular Weight (g/mol) 263.42
MDL Number MFCD00013045
InChI Key CNWGLQAFFSLHRX-UHFFFAOYSA-N
Synonym thallium i acetate
PubChem CID 131675083
IUPAC Name acetic acid;molecular hydrogen;thallium
SMILES [HH].CC(=O)O.[Tl]

Specifications

Melting Point 128.0°C to 130.0°C
Color White
Physical Form Fine Crystalline Powder
Quantity 25 g
Assay Percent Range 99%
Linear Formula CH3COOTl
Merck Index 15, 9405
Formula Weight 263.42
Percent Purity 99%
Infrared Spectrum Authentic
Packaging Glass bottle
Chemical Name or Material Thallous acetate
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0468042 Certificate TypeCertificate of Analysis Date31/01/2025 Catalog Number420771000,420775000,420770250
Lot NumberA0465547 Certificate TypeCertificate of Analysis Date20/08/2024 Catalog Number420771000,420770250,420775000
Lot NumberA0459650 Certificate TypeCertificate of Analysis Date25/01/2024 Catalog Number420771000,420770250,420775000
Lot NumberA0401256 Certificate TypeCertificate of Analysis Date14/12/2022 Catalog Number420771000,420770250,420775000
Lot NumberA0398223 Certificate TypeCertificate of Analysis Date14/12/2022 Catalog Number420771000,420770250,420775000
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Thallous acetate
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 1
  • Acute toxicity Category 2
  • LONG-TERM AQUATIC HAZARD Chronic 2
  • Specific target organ toxicity after repeated exposure Category 2
Hazard Statement
  • H310-Fatal in contact with skin.
  • H330-Fatal if inhaled.
  • H373-May cause damage to organs.
  • H411-Toxic to aquatic life with long lasting effects.
  • H300-Fatal if swallowed.
Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Thallous Acetate, 99%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.