Learn More
Abeba™ The Original 7030 Shoes
Animal fats, oil and fuel resistant. Sanitized™ treated.
Brand:  Abeba™ 7030/40
 View more versions of this product
Product Code. 15652488
  • 56.85 € / 1 pair
Estimated Shipment: 05-12-2024
to see stock.
Explore more special offers
Add to basket
Description
Description
  • CE EN ISO 20347:2012 OB FO SRC
  • Smooth leather, perforated
  • Black
  • Slip resistance outsole sole
  • Sanitized treated
  • Heel strap foldable, adjustable
  • Leather insole
Specifications
Specifications
CE EN ISO 20347:2012 OB FO SRC
Leather
Slip-resistant
Animal fats, oil and fuel
Black
40
7030