Learn More
GA International Thermal-Transfer Cryogenic Labels for Frozen Vials and Surfaces
Thermal-transfer cryogenic labels that will firmly adhere to both ambient and already frozen surfaces prior to long-term storage in liquid nitrogen.
Brand:  GA International UC-8C3-4WH
Encompass_Preferred
 View more versions of this product
Product Code. 17801298
  • POA / Pack of 4000
This item requires a price update prior to being ordered.
Request a Price for this item here.
View alternative products »
Explore more special offers
Description
Description
  • Ultra-resistant adhesive ensures no flagging or detachment when stored in liquid nitrogen
  • Thermal-transfer printable for the best printout and barcode quality
  • Ideal for labeling ambient temperature as well as frozen surfaces without thawing first
  • No overlap or wrap-around required for proper adhesion
  • Wide temperature range resistance
Specifications
Specifications
Permanent
Medium Gloss
Thermoplastic Film
Labels
2 Labels
Rectangle
-196°C to +121°C
0.5 in.
1 in.
3 in.
White
Roll
Thermal Transfer
Non-sterile
4000/Pk.
-321°F to +250°F
25.4 x 12.7 mm
12.7 mm
25.4 mm
76.2 mm
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
One moment while we fetch your results.
Certificates
Special Offers
Special Offers
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
GA International Thermal-Transfer Cryogenic Labels for Frozen Vials and Surfaces