missing translation for 'onlineSavingsMsg'
Learn More

Brady™ Thermal Transfer Printer Labels

Polypropylene Thermal Transfer Printer Labels

Brand:  Brady™ 302987

 View more versions of this product

Product Code. 17130481

  • 188.00 € / Pack of 1500
Estimated Shipment: 21-02-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Laboratory Applications:
    • Centrifuge
    • Eppendorf & PCR Tubes
    • Conical & Large Tubes
    • Vials & Tubes
  • Printer compatibility:
    • BBP™11 Label Printer
    • BBP™12 Label Printer
    • Brady IP™ Series Label Printer
    • BradyPrinter i5100 Industrial Label Printer
    • BradyPrinter i7100 Industrial Label Printer
    • THT 76 mm Core Printers
Specifications

Specifications

THT-309-7600-1.5SC
Translucent
Roll
Front
Labels
Label 0.5 - 8.0 mL Vial/Tube Diameter 11.18 mm
-196°C
-321°F to +248°F
0.076 mm
0.5 in.
1 in.
12.70 mm
300 g
Permanent Acrylic
Gloss
Polypropylene
Thermal Transfer
Non-sterile
BBP™11 Label Printer; BBP™12 Label Printer; THT 76 mm Core Printers
1500 Labels/Roll
-196°C to +120°C
15.24 mm
12.70 mm
25.40 mm
11.18 mm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.