Learn More
Thermographic Measurements Thermax 10-Level Temperature Indicator Strips 77°C to 127°C (171°F to 261°F)
Single use, irreversible threshold indicator reports the highest temperature reached.
Brand:  Thermographic Measurements M10STHERGBD01P1
Product Code. 17158244
  • 19.70 € / Pack of 10
In Stock
to see stock.
Edge
Explore more special offers
Add to basket
Description
Description
  • Immediate response
  • Permanent record of highest temperature achieved
  • Oil, water, and steam resistant
  • Inexpensive and easy to use
  • Self-adhesive
Specifications
Show More
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Thermographic Measurements Thermax 10-Level Temperature Indicator Strips 77°C to 127°C (171°F to 261°F) > Temperature Range: 77°C to 127°C (171°F to 261°F)