Learn More
Thermo Scientific™ 4-Place Swinging Bucket Rotor Adapters
For added processing versatility
Brand:  Thermo Scientific™ 75007542
 View more versions of this product
Product Code. 10759612
  • 74.57 € / Each
Explore more special offers
Description
Description
For added processing versatility
  • Maximum tube size: 30 x 117mm
Order Info
Order 4-Place Swinging Bucket Rotor (75-007-591) with Round Buckets (75-007-555). Adapters are sold individually; order quantity to equip rotor. Note: 50mL conical adapter (75-007-556) requires rubber pads (supplied with buckets). Remove pads when using all other size adapters.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
50 mL Tube
Swinging bucket rotor