Learn More
Thermo Scientific™ 4-Place Swinging Bucket Rotor Adapters
For added processing versatility
68.64 € - 120.60 €
Product Code | Brand | Tube Type | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Tube Type | Price | Quantity & Availability | |||||
10759612
|
Thermo Scientific™
75007542 |
50 mL Tube |
74.57 €
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
10294434
|
Thermo Scientific™
75007545 |
7 mL Tube |
70.65 €
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
10698802
|
Thermo Scientific™
75007544 |
15 mL Blood Tube |
73.06 €
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
10076261
|
Thermo Scientific™
75007547 |
Microtube |
120.60 €
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
10688802
|
Thermo Scientific™
75007557 |
Falcon |
68.64 €
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
10699762
|
Thermo Scientific™
75007556 |
50 mL Falcon Tube |
72.96 €
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
For added processing versatility
- Maximum tube size: 30 x 117mm
Order Info
Order 4-Place Swinging Bucket Rotor (75-007-591) with Round Buckets (75-007-555). Adapters are sold individually; order quantity to equip rotor. Note: 50mL conical adapter (75-007-556) requires rubber pads (supplied with buckets). Remove pads when using all other size adapters.
Specifications
Swinging bucket rotor |