missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Luminaris HiGreen qPCR Master Mix, low ROX

Thermo Scientific Luminaris Color HiGreen and Luminaris HiGreen qPCR Master Mixes are universal ready-to-use solutions optimized for qPCR and two-step RT-qPCR.

Brand:  Thermo Scientific™ K0971

136.00 EUR valid until 2024-12-31
Use promo code "21651" to get your promotional price.


 View more versions of this product

Product Code. 13585250

  • 272.00 € / 250 reactions

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers

Includes: Luminaris HiGreen Low ROX qPCR Master Mix (2X) containing Hot Start Taq DNA Polymerase, UDG, dNTPs (with dUTP) and SYBR Green I in an optimized PCR buffer. The master mix contains low concentration of ROX passive reference dye for specific real-ti
This item is not returnable. View return policy

Description

Description

  • UDG in the master mix to prevent carry-over contamination
  • Equal performance of blue and colorless master mixes
  • Versions with premixed ROX or fluorescein for different real-time platforms
  • Standard cycling protocols
  • Excellent qPCR performance
  • Specificity: Hot Start Taq DNA Polymerase and the optimized buffer eliminate non-specific amplification and formation of primer dimers
  • Sensitivity: single copy detection
  • Wide linear range: accurate quantification across 9 orders of magnitude
  • Reproducibility and convenience: ready-to-use 2X master mix
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Contains:
• 2 x 1.25 mL Luminaris HiGreen Low ROX qPCR Master Mix (2X) containing Hot Start Taq DNA Polymerase, UDG, dNTPs (with dUTP) and SYBR Green I in an optimized PCR buffer; sufficient for 250 x 20 μL reactions. The master mix contains low concentration of ROX passive reference dye for specific real-time platforms.
• 2 x 1.25 mL Nuclease-Free Water

Store at -20°C.
High
Taq DNA Polymerase
7500 System, QuantStudio™, ViiA™ 7 System, Stratagene Mx4000, Stratagene Mx3000P, Stratagene Mx3005P
qPCR Master Mix
250 Reactions
2X
250 Reactions
SYBR
qPCR
Standard
Luminaris
250 reactions
Gene Expression
2 X
DNA, dsDNA
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only. Not for use in diagnostic procedures.