Learn More
Thermo Scientific™ H-6000A Bucket Adapters
Accept a variety of blood processing tubes, bags and bottles
Brand: Thermo Scientific™ 00447
This item is not returnable.
View return policy
Description
Accept a variety of blood processing tubes, bags and bottles
For use with Thermo Scientific H-6000A buckets.
- Adapters hold a maximum capacity of 6 × 500mL (bottles), 6 × 550mL (blood bags), and 132 × 7mL (tubes). Max. O.D. × L: 70 x 160mm
Specifications
48 x 15 mL Conical Blood Collection Tube | |
H-6000A bucket for Thermo Scientific™ Sorvall™ RC3BP™ Plus low speed centrifuge rotor |
Product Content Correction
Product Title
Thermo Scientific™ H-6000A Bucket Adapters > Conical Blood Collection Tube; 15mL
Spot an opportunity for improvement?Share a Content Correction