Learn More
Thermo Scientific™ H-6000A Bucket Adapters
Accept a variety of blood processing tubes, bags and bottles
231.15 € - 806.01 €
Specifications
For Use With (Equipment) | H-6000A bucket for Thermo Scientific™ Sorvall™ RC3BP™ Plus low speed centrifuge rotor |
---|
Product Code | Brand | Capacity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Capacity | Price | Quantity & Availability | |||||
12843412
|
Thermo Scientific™
00384 |
132 x 7 mL Blood Collection Tubes |
717.57 €
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
12833412
|
Thermo Scientific™
00385 |
108 x 15 mL Blood Collection Tube |
273.36 €
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
10770414
|
Thermo Scientific™
00436 |
30 x 50 mL Tubes |
231.15 €
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
13144299
|
Thermo Scientific™
00444 |
6 x 500 mL Bottles |
430.14 €
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
12823412
|
Thermo Scientific™
00445 |
18 x 50 mL Tubes |
232.16 €
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
10242723
|
Thermo Scientific™
00447 |
48 x 15 mL Conical Blood Collection Tube |
344.72 €
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
12716967
|
Thermo Scientific™
00512 |
6 x 550 mL Blood Bags |
275.37 €
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
12853412
|
Thermo Scientific™
00857 |
5 mL Blood Collection Tubes |
806.01 €
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15129485
|
Thermo Scientific™
01092 |
6 x 250 mL Bottles |
232.16 €
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Accept a variety of blood processing tubes, bags and bottles
For use with Thermo Scientific H-6000A buckets.
- Adapters hold a maximum capacity of 6 × 500mL (bottles), 6 × 550mL (blood bags), and 132 × 7mL (tubes). Max. O.D. × L: 70 x 160mm
Specifications
H-6000A bucket for Thermo Scientific™ Sorvall™ RC3BP™ Plus low speed centrifuge rotor |