missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Pierce™ Triple Quadrupole Calibration Solution, extended mass range

Obtain both positive and negative ionization calibration of Thermo Scientific™ Triple Stage Quadrupole instruments using the Thermo Scientific™ Pierce™ Triple Quadrupole Calibration Solution, Extended Mass Range, which is a mixture of 14 highly pure, ionizable components (mass range: 69 m/z to 2800m/z).

Brand:  Thermo Scientific™ 88340

400.90 EUR valid until 2025-05-31
Use promo code "25977" to get your promotional price.


Product Code. 15322627

  • 427.00 € / 10mL

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

The Thermo Scientific Pierce Triple Quadrupole Calibration Solution, Extended Mass Range, is a mixture of 14 highly pure, ionizable components (mass range: 69 m/z to 2800m/z) designed for both positive and negative ionization calibration of Thermo Scientific Triple Stage Quadrupole instruments.

  • Improved mass accuracy - extended mass range designed to improve mass accuracy when compared to existing calibration solutions
  • Positive and negative mode calibration - single formulation with extended mass ranges for both ionization calibration modes
  • Highly validated - high-quality, rigorously tested formulation with lot-specific certificates of analysis
  • Ready to use - reference standard may be directly loaded into a syringe and injected into the instrument
  • Stable - store at 2 to 8°C for 18 months
The Pierce Triple Quadrupole Calibration Solution is a ready-to-use formulation that has been optimized for both positive and negative ionization calibration of instruments. The extended range masses (69 m/z to 2800m/z) improve performance of the Thermo Scientific Quantiva™ and Endura™ Triple Quadrupole mass spectrometers. Additionally, the Triple Quadrupole Calibration Solution, Extended Mass Range, is manufactured at an ISO 9001 facility and each lot is quality controlled with strict specifications. The stable solution is provided in a leak-proof, high-purity PTFE bottle.
The Pierce Triple Quadrupole Calibration Solution, Extended Mass Range, formulation contains 14 compounds that are ionizable in both positive and negative modes for instrument calibration. The formulation has been specifically optimized for use with the TSQ Endura™ Series and TSQ Quantiva™ Series instruments. The calibration solution is also compatible with the TSQ Quantum™ Series#, TSQ Discovery™ Series#, TSQ Quantum Ultra™ Series#, TSQ Quantum Access™ Series#, and TSQ Vantage™ Series# instruments using the list of recommended masses.
Positive mode (m/Z): 69.0447, 142.1590, 322.0481, 622.0289, 922.0098
Negative mode (m/Z): 59.0128, 112.9845, 301.9970, 601.9779, 1033.9870
For the calibration procedure use:
TSQ Series Getting Started Guide (70111-97152)

TRUSTED_SUSTAINABILITY
Specifications

Specifications

Mass Spectrometer
Pierce™
Peptides, Small Molecules
Mass Spectrometry
10 mL
Calibration Solution
1 x 10 mL
Store in refrigerator (2–8°C).
Peptides, Small Molecules
Calibration
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberZL401366 Certificate TypeCertificate of Analysis Date25/01/2025 Catalog Number88340
Lot NumberZJ399078 Certificate TypeCertificate of Analysis Date16/12/2024 Catalog Number88340
Lot NumberZJ399079 Certificate TypeCertificate of Analysis Date16/12/2024 Catalog Number88340
Lot NumberZJ399077 Certificate TypeCertificate of Analysis Date26/10/2024 Catalog Number88340
Lot NumberZI399076 Certificate TypeCertificate of Analysis Date15/10/2024 Catalog Number88340
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Thermo Scientific™ Pierce™ Triple Quadrupole Calibration Solution, extended mass range > 10mL

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only. Not for use in diagnostic procedures.