Learn More
Thermo Scientific™ Remel™ Burkholderia cepacia ATCC™ 25416, Culti-Loops™
Thermo Scientific Remel Burkholderia cepacia ATCC 25416, Culti-Loops, ATCC: 25416TM, Species: Burkholderia cepacia, Recommended Applications: Healthcare
Brand: Thermo Scientific™ R4605220
Description
Thermo Scientific Remel Burkholderia cepacia ATCC 25416, Culti-Loops, ATCC: 25416TM, Species: Burkholderia cepacia, Recommended Applications: Healthcare
The loop may be dissolved in liquid medium, Culti-Loop Aerobic Set Rehydration Fluid, Part No. R4631000 or streaked directly onto an appropriate media. Culti-Loops are recommended for use in performance testing of media, stains, reagents, and identification kits, and for the evaluation of bacteriological procedures. MycoBacti-Loops are ready to use, disposable inoculating loops containing stabilized, preserved, viable microorganisms.
Order Info
Culti-Loop products are individually sealed in a foil pouch and packaged five loops per package. Each Mycobacti-Loop is individually sealed in a foil pouch and packaged one loop per package. BactiDisk products are packaged 10 disks per vial.
Specifications
Burkholderia cepacia | |
Culti-Loop | |
UCB 717 [717-ICPB 25, Ballard 717, CCUG 12691, CCUG 13226, CFBP 2227, CIP 80.24, DSM 7288, HAMBI 1976, ICMP 5796, IFO 14074, JCM 5964, LMG 1222, NCCB 76047, NCPPB 2993, NCTC 10743, NRRL B-14810] |
25416™* | |
10 Disk/Vial |
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
236815 | Certificate of Analysis | 18/03/2025 | R4605220 |
200635 | Certificate of Analysis | 18/12/2024 | R4605220 |
160412 | Certificate of Analysis | 05/09/2024 | R4605220 |
124245 | Certificate of Analysis | 11/06/2024 | R4605220 |
814510 | Certificate of Analysis | 16/04/2024 | R4605220 |
Culti-Loops are CE marked.