missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Epredia™ Shandon™ Pathcentre™ Tissue Processor Accessories
For use with the Epredia™ Shandon™ Tissue Processor
Brand: Epredia™ A75210193
This item is not returnable.
View return policy
Specifications
Storage Bottle | |
Tissue Processor | |
3 Pk. |