Learn More
Thermo Scientific™ Sorvall™ Adapters for Stratos™ Four-Place Swinging Bucket Rotor
For use with Thermo Scientific Sorvall Stratos Centrifuges in Four-Place Swinging Bucket Rotor 75-003-047 with Round Bucket 75-008-172
Brand:  Thermo Scientific™ 75008186
 View more versions of this product
Product Code. 10039500
  • 92.96 € / Each
Explore more special offers
Description
Description
  • Accommodate tube sizes from 1.5 to 100mL, including urine tubes
Order Info
Adapters are sold individually; order correct quantity to equip rotor.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
4 x 180 Swing bucket rotor
Red