Learn More
Thermo Scientific™ Sorvall™ Adapters for Stratos™ Four-Place Swinging Bucket Rotor
For use with Thermo Scientific Sorvall Stratos Centrifuges in Four-Place Swinging Bucket Rotor 75-003-047 with Round Bucket 75-008-172
90.35 € - 94.67 €
Specifications
For Use With (Equipment) 4 x 180 Swing bucket rotor
View More Specs
Products 6
Product Code Brand Color Price Quantity & Availability  
Product Code Brand Color Price Quantity & Availability  
10410083
View Documents
Thermo Scientific™
75008183
Green/Yellow
94.67 €
Each
10459502
View Documents
Thermo Scientific™
75008182
Green
92.96 €
Each
10358082
View Documents
Thermo Scientific™
75008187
Brown
93.47 €
Each
10509151
View Documents
Thermo Scientific™
75008185
White
93.47 €
Each
10039500
View Documents
Thermo Scientific™
75008186
Red
92.96 €
Each
10568761
View Documents
Thermo Scientific™
Gray This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
Description
Description
  • Accommodate tube sizes from 1.5 to 100mL, including urine tubes
Order Info
Adapters are sold individually; order correct quantity to equip rotor.
Specifications
Specifications
4 x 180 Swing bucket rotor