missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Tips for Tecan™ and Axon™ Instruments PatchXpress Workstations
GREENER_CHOICE

For use with Tecan™ Genesis™, Freedom Evo™, Cavro™, PatchXpress Workstations

133.00 € - 140.00 €

Specifications

Volume (Metric) 50 μL
Filtered Filtered
Color White
Autoclavable Autoclavable
Clean Claims DNase, RNase, and Pyrogen free
View More Specs

Products 2
Product Code Brand Sterility Price Quantity & Availability  
Product Code Brand Sterility Price Quantity & Availability  
11510205
GREENER_CHOICE
View Documents
Thermo Scientific™
902-262XP
Non-sterile
133.00 €
Pack of 960
Estimated Shipment: 25-06-2025
to see stock.
Add to basket
 
11500205
GREENER_CHOICE
View Documents
Thermo Scientific™
902-261XP
Sterile
140.00 €
Pack of 960
Estimated Shipment: 14-07-2025
to see stock.
Add to basket
 
Description

Description

For use with Tecan™ Genesis™, Freedom Evo™, Cavro™, PatchXpress Workstations

Clear pipet tips, Tecan style.

Specifications

Specifications

50 μL
White
DNase, RNase, and Pyrogen free
Automation
Polypropylene
Filtered
Autoclavable
Rack
Tecan Genesis, Tecan Freedom EVO, Cavro, patchXpress
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.