Learn More
Sonoco™ ThermoSafe LabMailer Specimen Tube Mailers
Shock-absorbing polystyrene foam mailers accept sample tubes with rubber or screw caps
Brand:  Sonoco™ 409
 View more versions of this product
Product Code. 17808703
  • 203.00 € / Pack of 100
Estimated Shipment: 04-07-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Two-piece assembly with snap-lock fasteners holds contents in place
  • Ideal for mailing or transporting nonhazardous blood, urine, or other lab samples.
†Includes slotted area for up to four slides.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
5 Tubes
7.875 x 5.125 x 1.5 in.
170 mm
7, 10, 15 or 25 mL
Ideal for mailing or transporting nonhazardous blood, urine, or other lab samples
16 mm
Foam only shipper. Must purchase mailing sleeve model 410, Holds 5 Tubes
Polystyrene Foam
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Sonoco™ ThermoSafe LabMailer Specimen Tube Mailers > Tube Cavity: 105 x 17mm; No. of Tubes: 5