missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Product Code | Brand | Dimensions (LxWxH) | Weight (English) | Weight (Metric) | Price | Quantity & Availability | ||||||
---|---|---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Dimensions (LxWxH) | Weight (English) | Weight (Metric) | Price | Quantity & Availability | ||||||
13059342
|
Sonoco™
FPP18 |
8.25 x 10 x 0.75 in. (20.9 x 20.3 x 1.9 cm) | 18 oz. | 0.72 kg |
64.00 €
Pack of 28 |
Estimated Shipment: 20-06-2025
Log in to see stock. |
||||||
13079342
|
Sonoco™
FPP24 |
6.5 x 4.5 x 1.5 in. (16.51 x 11.43 x 3.81 cm) | 24 oz. | 0.68 kg |
52.00 €
Pack of 24 |
Estimated Shipment: 02-07-2025
Log in to see stock. |
||||||
13089342
|
Sonoco™
FPP31 |
9 x 4 x 1.5 in. (22.8 x 10.16 x 3.81 cm) | 31 oz. | 1.2 kg | N/A | N/A | N/A | |||||
13039342
|
Sonoco™
FPP7B |
4.5 x 4 x 0.75 in. (11.4 x 53.34 x 1.9 cm) | 7.5 oz. | 0.21 kg | N/A | N/A | N/A | |||||
Description
Refrigerant bricks are vacuum-sealed for consistent weight and shape. These semi-rigid foam bricks never change shape during freezing, thawing or transit, enabling superior surface contact for more consistent temperature control.
- Available in a broad selection of styles and sizes
- Maintain temperature for an extended time
- Never change shape while in transit
- Freeze at 0°C (32°F)
- Nontoxic
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Sonoco™ ThermoSafe PolarPack™ Foam Bricks