missing translation for 'onlineSavingsMsg'
Learn More
Thiazolyl blue tetrazolium bromide, 98%, Thermo Scientific Chemicals Product Code.: 15214654

Thiazolyl blue tetrazolium bromide, 98%, Thermo Scientific Chemicals

Product Code. 15214654
1 g, Glass bottle
Click to view available options
Quantity:
250 mg
1 g
5 g
Packaging:
Glass bottle
This item is not returnable. View return policy

Product Code. 15214654

Brand: Thermo Scientific Acros 158990010

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 298-93-1
Molecular Formula C18H16BrN5S
Molecular Weight (g/mol) 414.33
MDL Number MFCD00011964,MFCD00066662
InChI Key AZKSAVLVSZKNRD-UHFFFAOYSA-M
Synonym thiazolyl blue, thiazolyl blue tetrazolium bromide, mmt tetrazolium, methylthiazoletetrazolium, mtt, thiazolyl blue monotetrazolium, thiazole blue, 3-4,5-dimethylthiazol-2-yl-2,5-diphenyltetrazolium bromide, mtt van, unii-euy85h477iShow MoreShow Less
PubChem CID 64965
ChEBI CHEBI:53233
SMILES [Br-].CC1=C(C)N=C(S1)[N+]1=NC(=NN1C1=CC=CC=C1)C1=CC=CC=C1

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 195.0°C
Assay Percent Range 97.5% min. (Argentometry)
Packaging Glass bottle
Quantity 1 g
Beilstein 27,IV,6045
Solubility Information Solubility in water: 10g/L (20°C). Other solubilities: soluble in 5% methanol, soluble in ethanol
IUPAC Name 2-(3,5-diphenyltetrazol-2-ium-2-yl)-4,5-dimethyl-1,3-thiazole;bromide
Formula Weight 414.32
Percent Purity 98%
Chemical Name or Material Thiazolyl blue tetrazolium bromide, 98%
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0461943 Certificate TypeCertificate of Analysis Date19/04/2024 Catalog Number158990010,158992500,158990050
Lot NumberA0449546 Certificate TypeCertificate of Analysis Date20/01/2023 Catalog Number158990010,158992500,158990050
Lot NumberA0394701 Certificate TypeCertificate of Analysis Date15/12/2022 Catalog Number158990010,158992500,158990050
Lot NumberA0413402 Certificate TypeCertificate of Analysis Date13/12/2022 Catalog Number158990010,158992500,158990050
Lot NumberA0404305 Certificate TypeCertificate of Analysis Date13/12/2022 Catalog Number158990010,158992500,158990050
compliance-icons compliance-icons
Product Identifier
  • Thiazolyl blue tetrazolium bromide
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Germ cell mutagenicity Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H341-Suspected of causing genetic defects.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Thiazolyl blue tetrazolium bromide, 98%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.