missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Wilmad-LabGlass™ Thin Walled High Throughput NMR Tubes
Possess an average camber of 60 microns to guarantee the clarity of the spectrum for small molecule (MW<250) samples up to 600 MHz. Wilmad Thin Walled High Throughput NMR Tubes are designed for routine use in most low to mid field NMR spectrometers.
78.16 € - 290.73 €
Specifications
Camber (Metric) | 60 μm |
---|---|
Material | ASTM E438 Type 1 Class B Glass |
Product Type | NMR Tube |
Thickness (Metric) Wall | 0.43 mm |
Product Code | Brand | Length (Metric) | Field Strength | Quantity | Price | Quantity & Availability | ||||||
---|---|---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Length (Metric) | Field Strength | Quantity | Price | Quantity & Availability | ||||||
15398628
|
Wilmad-LabGlass™
66000-0061 |
103.5 mm | 600 MHz | 100/Pk. |
281.00 €
Pack of 100 |
Estimated Shipment: 30-05-2025
Log in to see stock. |
||||||
15309038
|
Wilmad-LabGlass™
66999-5700 |
178 mm | High-throughput | 100/Pk. |
180.00 €
Pack of 100 |
In Stock
Log in to see stock. ![]() |
||||||
15319038
|
Wilmad-LabGlass™
66999-5750 |
178 mm | High-throughput | 50/Pk. | N/A | N/A | N/A | |||||
15348698
|
Wilmad-LabGlass™
66000-0165 |
178 mm | 600 MHz | 100/Pk. |
290.73 €
Pack of 100 |
In Stock
Log in to see stock. |
||||||
15329038
|
Wilmad-LabGlass™
66999-5800 |
203 mm | High-throughput | 100/Pk. |
215.00 €
Pack of 100 |
Estimated Shipment: 12-06-2025
Log in to see stock. |
||||||
15339038
|
Wilmad-LabGlass™
66999-5850 |
203 mm | High-throughput | 50/Pk. | N/A | N/A | N/A | |||||
Description
- Camber and concentricity values listed are the Total Indicator Reading (TIR)
- Designed for routine use in most low to mid field NMR spectrometers
- One of the best OD tolerances in the industry
- Guaranteed to fit tightly in spinner turbine
- Made from ASTM E438 Type 1 Class B glass
- 100% inspected for surface defects and physical dimension to ensure the success of your experiments
- WG-1000-4-SJ and WG-1000-7-SJ are supplied with authentic Bruker SampleJet caps, all others come with a disposable NMR cap
Specifications
60 μm | |
ASTM E438 Type 1 Class B Glass | |
NMR Tube | |
0.43 mm |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Wilmad-LabGlass™ Thin Walled High Throughput NMR Tubes