Learn More
Thiourea, 99+%, ACS reagent, Thermo Scientific Chemicals
61.10 € - 170.00 €
Chemical Identifiers
CAS | 62-56-6 |
---|---|
Molecular Formula | CH4N2S |
Molecular Weight (g/mol) | 76.12 |
MDL Number | MFCD00008067 |
InChI Key | UMGDCJDMYOKAJW-UHFFFAOYSA-N |
Synonym | thiocarbamide, 2-thiourea, pseudothiourea, isothiourea, sulourea, 2-thiopseudourea, sulfourea, thiuronium, thiocarbonic acid diamide, urea, thio |
PubChem CID | 2723790 |
ChEBI | CHEBI:36946 |
IUPAC Name | thiourea |
SMILES | NC(N)=S |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10790682
|
Thermo Scientific Acros
424542500 |
250 g | Plastic bottle |
61.10 €
250g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10780682
|
Thermo Scientific Acros
424540010 |
1 kg | Plastic bottle |
170.00 €
1kg |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
62-56-6 | |
76.12 | |
UMGDCJDMYOKAJW-UHFFFAOYSA-N | |
2723790 | |
thiourea |
CH4N2S | |
MFCD00008067 | |
thiocarbamide, 2-thiourea, pseudothiourea, isothiourea, sulourea, 2-thiopseudourea, sulfourea, thiuronium, thiocarbonic acid diamide, urea, thio | |
CHEBI:36946 | |
NC(N)=S |
Specifications
62-56-6 | |
White | |
0.5% max. | |
99% min. (On dried basis) (Argentometry) | |
H2NCSNH2 | |
03, 180 | |
(to bismuth) Passes Test | |
thiocarbamide, 2-thiourea, pseudothiourea, isothiourea, sulourea, 2-thiopseudourea, sulfourea, thiuronium, thiocarbonic acid diamide, urea, thio | |
UMGDCJDMYOKAJW-UHFFFAOYSA-N | |
thiourea | |
2723790 | |
76.12 | |
ACS Reagent | |
Thiourea, 99+% |
171.0°C to 175.0°C | |
0.1% max. | |
Authentic | |
CH4N2S | |
MFCD00008067 | |
01,1164; 02,412; 03,290; 06,586; 11,519; 15,308 | |
15, 9521 | |
(16.5g in 100ml water,25°C) Passes test | |
NC(N)=S | |
76.12 | |
CHEBI:36946 | |
99+% | |
Crystalline Powder |
Safety and Handling
GHS H Statement
Suspected of causing cancer.
Toxic to aquatic life with long lasting effects.
Suspected of damaging the unborn child.
Harmful if swallowed.
GHS P Statement
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Immediately call a POISON CENTER or doctor/physician.
Use personal protective equipment as required.
IF INHALED: Remove victim to fresh air and keep
GHS Signal Word: Warning
missing translation for 'einecsNumber' : 200-543-5
missing translation for 'rtecsNumber' : YU2800000
missing translation for 'tsca' : TSCA
- Thiourea
Signal Word
- Warning
Hazard Category
- Acute toxicity Category 4
- Carcinogenicity Category 2
- Reproductive toxicity Category 2
- LONG-TERM AQUATIC HAZARD Chronic 2
Hazard Statement
- H351-Suspected of causing cancer.
- H411-Toxic to aquatic life with long lasting effects.
- H361d-Suspected of damaging the unborn child.
- H302-Harmful if swallowed.
Precautionary Statement
- P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
- P310-Immediately call a POISON CENTER/doctor/.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P273-Avoid release to the environment.
RUO – Research Use Only