Learn More
Idex Threaded Luer PEEK Adapter
Convenience of quick connect and disconnect of a luer connection
Brand:  Idex 02023-59
 View more versions of this product
Product Code. 12947759
  • 32.60 € / Each
Estimated Shipment: 08-01-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
Convenience of quick connect and disconnect of a luer connection
  • Manufactured from inert polymers
Quick connect luer adapters are designed to connect any female luer to any male luer in a variety of applications. Use with male luer lock adapters for more secure connection.
Specifications
Specifications
Threaded Luer Adapter
125°C
Black
0.05 in.
PEEK
Female Luer x Female M6 Flat Bottom