missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific Chemicals Thymidine, 99+%
162.00 € - 734.00 €
Identificadores de productos químicos
CAS | 50-89-5 |
---|---|
Molecular Formula | C10H14N2O5 |
Molecular Weight (g/mol) | 242.23 |
InChI Key | IQFYYKKMVGJFEH-UHFFFAOYNA-N |
Synonym | thymidine, deoxythymidine, 2'-deoxythymidine, 5-methyldeoxyuridine, thymidin, beta-thymidine, dthyd, 5-methyl-2'-deoxyuridine, thymine-2-deoxyriboside, thyminedeoxyriboside |
PubChem CID | 5789 |
ChEBI | CHEBI:17748 |
IUPAC Name | 1-[(2R,4S,5R)-4-hydroxy-5-(hydroxymethyl)oxolan-2-yl]-5-methylpyrimidine-2,4-dione |
SMILES | CC1=CN(C2CC(O)C(CO)O2)C(=O)NC1=O |
Código de producto | Marca | Quantity | Packaging | Precio | Cantidad y disponibilidad | ||||
---|---|---|---|---|---|---|---|---|---|
Código de producto | Marca | Quantity | Packaging | Precio | Cantidad y disponibilidad | ||||
10053050
|
Thermo Scientific Chemicals
226740050 |
5 g | Glass bottle |
162.00 €
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10033433
|
Thermo Scientific Chemicals
226740250 |
25 g | Glass bottle |
535.00 €
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
11986811
|
Thermo Scientific Chemicals
226741000 |
100 g | Glass bottle |
734.00 €
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Descripción
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Identificadores de productos químicos
50-89-5 | |
242.23 | |
thymidine, deoxythymidine, 2'-deoxythymidine, 5-methyldeoxyuridine, thymidin, beta-thymidine, dthyd, 5-methyl-2'-deoxyuridine, thymine-2-deoxyriboside, thyminedeoxyriboside | |
CHEBI:17748 | |
CC1=CN(C2CC(O)C(CO)O2)C(=O)NC1=O |
C10H14N2O5 | |
IQFYYKKMVGJFEH-UHFFFAOYNA-N | |
5789 | |
1-[(2R,4S,5R)-4-hydroxy-5-(hydroxymethyl)oxolan-2-yl]-5-methylpyrimidine-2,4-dione |
Especificaciones
50-89-5 | |
187.0°C to 189.0°C | |
Authentic | |
C10H14N2O5 | |
+ 18.50 (20.00°C c=3,H2O) | |
thymidine, deoxythymidine, 2'-deoxythymidine, 5-methyldeoxyuridine, thymidin, beta-thymidine, dthyd, 5-methyl-2'-deoxyuridine, thymine-2-deoxyriboside, thyminedeoxyriboside | |
98.0 to 102% (lambda max. 267nm,pH 7) molar absorptivity 9600+/-4%,on dry basis | |
CC1=CN(C2CC(O)C(CO)O2)C(=O)NC1=O | |
242.23 | |
CHEBI:17748 | |
99+% | |
Thymidine |
0.62 to 0.68 (A250/A260),0.70 to 0.76 (A280/A260) | |
White | |
99% min. (HPLC) | |
159552 | |
+ 18.50 | |
Solubility in water: 46g/L (20°C). Other solubilities: soluble in glacial acetic acid, soluble in methanol, hot ethanol, hot acetone, hot, ethyl acetate and pyridine, sparingly soluble in hot chloroform | |
IQFYYKKMVGJFEH-UHFFFAOYNA-N | |
1-[(2R,4S,5R)-4-hydroxy-5-(hydroxymethyl)oxolan-2-yl]-5-methylpyrimidine-2,4-dione | |
5789 | |
242.23 | |
Crystalline Powder |
Seguridad y manipulación
Warning
missing translation for 'einecsNumber' : 200-070-4
RUO – Research Use Only