Learn More
Thermo Scientific Chemicals Thymol Blue, Indicator
39.70 € - 343.71 €
Chemical Identifiers
CAS | 76-61-9 |
---|---|
Molecular Formula | C27H30O5S |
Molecular Weight (g/mol) | 466.592 |
MDL Number | MFCD00005869 |
InChI Key | PRZSXZWFJHEZBJ-UHFFFAOYSA-N |
Synonym | TB, Thymolsulfonephthalein |
PubChem CID | 65565 |
IUPAC Name | 4-[3-(4-hydroxy-2-methyl-5-propan-2-ylphenyl)-1,1-dioxo-2,1$l^{6}-benzoxathiol-3-yl]-5-methyl-2-propan-2-ylphenol |
SMILES | CC1=CC(=C(C=C1C2(C3=CC=CC=C3S(=O)(=O)O2)C4=CC(=C(C=C4C)O)C(C)C)C(C)C)O |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10474015
|
Thermo Scientific Chemicals
151450050 |
5 g | Glass bottle |
39.70 €
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10397510
|
Thermo Scientific Chemicals
151450250 |
25 g | Glass bottle |
113.57 €
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10574074
|
Thermo Scientific Chemicals
151451000 |
100 g | Glass bottle |
343.71 €
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
IndicatorChemical Identifiers
76-61-9 | |
466.592 | |
PRZSXZWFJHEZBJ-UHFFFAOYSA-N | |
65565 | |
CC1=CC(=C(C=C1C2(C3=CC=CC=C3S(=O)(=O)O2)C4=CC(=C(C=C4C)O)C(C)C)C(C)C)O |
C27H30O5S | |
MFCD00005869 | |
TB, Thymolsulfonephthalein | |
4-[3-(4-hydroxy-2-methyl-5-propan-2-ylphenyl)-1,1-dioxo-2,1$l^{6}-benzoxathiol-3-yl]-5-methyl-2-propan-2-ylphenol |
Specifications
221.0°C | |
C27H30O5S | |
15, 9555 | |
Solubility in water: insoluble. Other solubilities: soluble in diluted alkali solutions, soluble in alcohol | |
Authentic | |
4-[3-(4-hydroxy-2-methyl-5-propan-2-ylphenyl)-1,1-dioxo-2,1$l^{6}-benzoxathiol-3-yl]-5-methyl-2-propan-2-ylphenol | |
65565 | |
Pure | |
Acid range: from pH 1.2 (red) to pH 2.8 (yellow), alkaline range: from pH 8.0 (yellow) to pH 9.6 (blue) | |
Fine Crystalline Powder |
76-61-9 | |
MFCD00005869 | |
TB, Thymolsulfonephthalein | |
PRZSXZWFJHEZBJ-UHFFFAOYSA-N | |
CC1=CC(=C(C=C1C2(C3=CC=CC=C3S(=O)(=O)O2)C4=CC(=C(C=C4C)O)C(C)C)C(C)C)O | |
466.592 | |
466.58 | |
700 min. (596nm, in 0.01N NaOH) | |
Brown to Green | |
Thymol Blue |
Safety and Handling
missing translation for 'einecsNumber' : 200-973-3
missing translation for 'rtecsNumber' : XP2575000
missing translation for 'tsca' : TSCA
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.