missing translation for 'onlineSavingsMsg'
Learn More
Tin foil, 0.05mm (0.002in) thick, 99.999% (metals basis) Product Code.: 15446255

Tin foil, 0.05mm (0.002in) thick, 99.999% (metals basis)

Product Code. 15446255
50 x 100 mm
Click to view available options
Quantity:
50 x 100 mm
This item is not returnable. View return policy

Product Code. 15446255

Brand: Thermo Scientific Alfa Aesar 038538.FY

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 7440-31-5 | Sn | 118.71 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

CAS 7440-31-5
Molecular Formula Sn
MDL Number MFCD00133862
Quantity 50 x 100 mm
Synonym powder, stannum, metallic, tin, elemental, wang, zinn, flake, tin, metal, silver matt powder, zinn german
InChI Key ATJFFYVFTNAWJD-UHFFFAOYSA-N
SMILES [Sn]
IUPAC Name tin
Molecular Weight (g/mol) 118.71
PubChem CID 5352426
ChEBI CHEBI:32990
Percent Purity 99.999%
Odor Odorless
Weight ≈1.8g/50x100mm
Form Foil
Assay (metals basis)
Chemical Name or Material Tin foil, 0.05mm (0.002 in.) thick
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberX17K045 Certificate TypeCertificate of Analysis Date17/12/2024 Catalog Number038538.FY
Lot NumberQ11K033 Certificate TypeCertificate of Analysis Date11/06/2024 Catalog Number038538.FY
Lot NumberU01J049 Certificate TypeCertificate of Analysis Date11/06/2024 Catalog Number038538.FY
Lot NumberQ14I041 Certificate TypeCertificate of Analysis Date24/05/2022 Catalog Number038538.FY
Lot NumberK22R024 Certificate TypeCertificate of Analysis Date08/12/2021 Catalog Number038538.FY

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Tin foil, 0.05mm (0.002in) thick, 99.999% (metals basis) >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.