missing translation for 'onlineSavingsMsg'
Learn More
Tin wire, 1.0mm (0.04in) dia, Puratronic™, 99.998% (metals basis), Thermo Scientific Chemicals Product Code.: 11366358

Tin wire, 1.0mm (0.04in) dia, Puratronic™, 99.998% (metals basis), Thermo Scientific Chemicals

Product Code. 11366358
50 cm
Click to view available options
Quantity:
50 cm
250 cm
1000 cm
This item is not returnable. View return policy

Product Code. 11366358

Brand: Thermo Scientific Alfa Aesar 011905.BW

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Form: approx. 5.72g/m

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7440-31-5
Molecular Formula Sn
Molecular Weight (g/mol) 118.71
MDL Number MFCD00133862
InChI Key ATJFFYVFTNAWJD-UHFFFAOYSA-N
Synonym powder, stannum, metallic, tin, elemental, wang, zinn, flake, tin, metal, silver matt powder, zinn german
PubChem CID 5352426
ChEBI CHEBI:32990
IUPAC Name tin
SMILES [Sn]

Specifications

Color Silver to Gray
Appearance Silver Gray wire
Density 7.31g/cm3
Assay Percent Range 99.998% (Metals basis)
Quantity 50 cm
Odor Odorless
Form Wire
Chemical Name or Material Tin

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberZ20K037 Certificate TypeCertificate of Analysis Date06/01/2025 Catalog Number011905.CD,011905.BW,011905.CB
Lot NumberW10K006 Certificate TypeCertificate of Analysis Date02/10/2024 Catalog Number011905.CD,011905.BW,011905.CB
Lot NumberW10K007 Certificate TypeCertificate of Analysis Date27/09/2024 Catalog Number011905.CD,011905.BW,011905.CB
Lot NumberG17R018 Certificate TypeCertificate of Analysis Date08/12/2021 Catalog Number011905.CD,011905.BW,011905.CB
Lot NumberQ16E014 Certificate TypeCertificate of Analysis Date08/12/2021 Catalog Number011905.CD,011905.BW,011905.CB

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Tin wire, 1.0mm (0.04in) dia, Puratronic™, 99.998% (metals basis), Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.