missing translation for 'onlineSavingsMsg'
Learn More

Tin(II) 2-ethylhexanoate, tech., Thermo Scientific Chemicals

45.23 € - 314.57 €

Chemical Identifiers

CAS 301-10-0
Molecular Formula C16H30O4Sn
Molecular Weight (g/mol) 405.122
MDL Number MFCD00002676
InChI Key KSBAEPSJVUENNK-UHFFFAOYSA-L
Synonym stannous octoate, tin ii 2-ethylhexanoate, tin octoate, tin ii bis 2-ethylhexanoate, tin 2+ bis 2-ethylhexanoate, tin ethylhexanoate, tin 2-ethylhexanoate, hexanoic acid, 2-ethyl-, tin 2+ salt, stannous 2-ethylhexoate, tin ii 2-ethylhexylate
PubChem CID 9318
IUPAC Name 2-ethylhexanoate;tin(2+)
SMILES CCCCC(CC)C(=O)[O-].CCCCC(CC)C(=O)[O-].[Sn+2]
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11333447
View Documents
Thermo Scientific Alfa Aesar
018590.22
100 g
45.23 €
100g
Please to purchase this item. Need a web account? Register with us today!
11343447
View Documents
Thermo Scientific Alfa Aesar
018590.36
500 g
120.60 €
500g
Please to purchase this item. Need a web account? Register with us today!
11323447
View Documents
Thermo Scientific Alfa Aesar
018590.A3
2 kg
314.57 €
2kg
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Tin(II) 2-ethylhexanoate is used as a polymerization initiator in polylactic acid production. It acts as an intermediate as well as a catalyst for urethane foams, lubricants, addition agents and stabilizers for transformer oils.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

301-10-0
405.122
KSBAEPSJVUENNK-UHFFFAOYSA-L
9318
CCCCC(CC)C(=O)[O-].CCCCC(CC)C(=O)[O-].[Sn+2]
C16H30O4Sn
MFCD00002676
stannous octoate, tin ii 2-ethylhexanoate, tin octoate, tin ii bis 2-ethylhexanoate, tin 2+ bis 2-ethylhexanoate, tin ethylhexanoate, tin 2-ethylhexanoate, hexanoic acid, 2-ethyl-, tin 2+ salt, stannous 2-ethylhexoate, tin ii 2-ethylhexylate
2-ethylhexanoate;tin(2+)

Specifications

301-10-0
1.251 g/mL
C16H30O4Sn
MFCD00002676
Miscible with water.
CCCCC(CC)C(=O)[O-].CCCCC(CC)C(=O)[O-].[Sn+2]
405.122
405.11
Liquid
Colorless to Yellow
>110°C (230°F)
1.494
stannous octoate, tin ii 2-ethylhexanoate, tin octoate, tin ii bis 2-ethylhexanoate, tin 2+ bis 2-ethylhexanoate, tin ethylhexanoate, tin 2-ethylhexanoate, hexanoic acid, 2-ethyl-, tin 2+ salt, stannous 2-ethylhexoate, tin ii 2-ethylhexylate
KSBAEPSJVUENNK-UHFFFAOYSA-L
2-ethylhexanoate;tin(2+)
9318
Technical
Tin(II) 2-ethylhexanoate
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H361-H315-H319-H335
Suspected of damaging fertility or the unborn child.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P201-P202-P261-P264b-P271-P280i-P281-P302+P352-P304+P340-P305+P351+P338-P308+P313-P332+P313-P362-P501c

H315-H319-H335-H361d

missing translation for 'einecsNumber' : 206-108-6

missing translation for 'rtecsNumber' : MO7870000

missing translation for 'tsca' : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.