Learn More
Tin(II) trifluoromethanesulfonate, 97%, Thermo Scientific Chemicals
49.95 € - 200.00 €
Chemical Identifiers
CAS | 62086-04-8 |
---|---|
Molecular Formula | CH4F3O3SSn+ |
Molecular Weight (g/mol) | 271.805 |
MDL Number | MFCD00191251 |
InChI Key | IACSYIHNPIQPMP-UHFFFAOYSA-M |
Synonym | tin ii trifluoromethanesulfonate |
PubChem CID | 131676109 |
IUPAC Name | molecular hydrogen;tin(2+);trifluoromethanesulfonate |
SMILES | [HH].[HH].C(F)(F)(F)S(=O)(=O)[O-].[Sn+2] |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11384527
|
Thermo Scientific Alfa Aesar
039603.03 |
1 g |
49.95 €
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11394527
|
Thermo Scientific Alfa Aesar
039603.06 |
5 g |
200.00 €
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Tin(II) trifluoromethanesulfonate is a mild Lewis acid used for stereoselective aldol and Michael reactions; [2,3]-Wittig and Ireland-Claisen rearrangements, Horner-Wadsworth-Emmons reactions, asymmetric [3 + 2] cycloaddition, ring opening reactions and Friedel-Crafts reactions.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
62086-04-8 | |
271.805 | |
IACSYIHNPIQPMP-UHFFFAOYSA-M | |
131676109 | |
[HH].[HH].C(F)(F)(F)S(=O)(=O)[O-].[Sn+2] |
CH4F3O3SSn+ | |
MFCD00191251 | |
tin ii trifluoromethanesulfonate | |
molecular hydrogen;tin(2+);trifluoromethanesulfonate |
Specifications
62086-04-8 | |
CH4F3O3SSn+ | |
UN3288 | |
tin ii trifluoromethanesulfonate | |
IACSYIHNPIQPMP-UHFFFAOYSA-M | |
molecular hydrogen;tin(2+);trifluoromethanesulfonate | |
131676109 | |
97% | |
Tin(II) trifluoromethanesulfonate |
(decomposition) | |
MFCD00191251 | |
Moisture sensitive | |
Insoluble in water. | |
[HH].[HH].C(F)(F)(F)S(=O)(=O)[O-].[Sn+2] | |
271.805 | |
416.82 | |
Powder |
Safety and Handling
GHS H Statement
H331-H302-H312
Toxic if inhaled.
Harmful if swallowed.
Harmful in contact with skin.
P260-P264b-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
H314-H500
missing translation for 'dotInformation' : Hazard Class: 8; Packaging Group: III
missing translation for 'tsca' : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.