Learn More
Tin(IV) iodide, 95%, Thermo Scientific Chemicals
69.70 € - 253.00 €
Chemical Identifiers
CAS | 7790-47-8 |
---|---|
Molecular Formula | H4I4Sn |
Molecular Weight (g/mol) | 630.36 |
MDL Number | MFCD00036272 |
InChI Key | ZBCVLXKKLMNLGV-UHFFFAOYSA-N |
Synonym | stannic iodide, stannane, tetraiodo, tin iv iodide, tetraiodotin, tin tetraiodide, tin iodide, unii-8h688bw69x, tin iv iodide 1:4, tin-iv-iodide |
PubChem CID | 24631 |
IUPAC Name | tetraiodostannane |
SMILES | [Sn].I.I.I.I |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11324809
|
Thermo Scientific Alfa Aesar
071114.09 |
10 g |
69.70 €
10g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11334809
|
Thermo Scientific Alfa Aesar
071114.18 |
50 g |
253.00 €
50g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Used in chemical research and as a food additive.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
7790-47-8 | |
630.36 | |
ZBCVLXKKLMNLGV-UHFFFAOYSA-N | |
24631 | |
[Sn].I.I.I.I |
H4I4Sn | |
MFCD00036272 | |
stannic iodide, stannane, tetraiodo, tin iv iodide, tetraiodotin, tin tetraiodide, tin iodide, unii-8h688bw69x, tin iv iodide 1:4, tin-iv-iodide | |
tetraiodostannane |
Specifications
340°C | |
7790-47-8 | |
Crystalline | |
SnI4 | |
UN3260 | |
stannic iodide, stannane, tetraiodo, tin iv iodide, tetraiodotin, tin tetraiodide, tin iodide, unii-8h688bw69x, tin iv iodide 1:4, tin-iv-iodide | |
ZBCVLXKKLMNLGV-UHFFFAOYSA-N | |
tetraiodostannane | |
24631 | |
95% | |
Moisture sensitive | |
Tin (IV) iodide |
∼143°C | |
Red | |
H4I4Sn | |
MFCD00036272 | |
14,8777 | |
Soluble in alcohol,benzene,CS2,chloroform,and ether; Decomposes in water | |
[Sn].I.I.I.I | |
630.36 | |
626.31 | |
2.106 | |
4.46 g/mL |
Safety and Handling
GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
H314-H335
missing translation for 'dotInformation' : Hazard Class: 8; Packaging Group: II
missing translation for 'einecsNumber' : 232-208-4
missing translation for 'rtecsNumber' : XQ3675000
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only