Learn More
Epredia™ TissueLoc™ Cassette IV Slotted Cassettes
For routine tissue samples, cassettes have a back-edge lid hinge with loose-packed lids that are available in 12 different color varieties
Brand:  Epredia™ B851954GN
102.00 EUR valid until 2024-12-31
Use promo code "21628" to get your promotional price.
 View more versions of this product
Product Code. 16324176
  • 140.00 € / Pack of 1000
Explore more special offers
Description
Description
Printing
  • Surface finish and reinforced print faces are engineered to optimize print quality
  • Tube design is precisely engineered to reduce jamming and prevent downtime
Grossing, Processing and Embedding
  • Cassette design and flow characteristics tested to ensure efficient reagent exchange
  • Opening and closing forces adjusted for ease of use, while safeguarding valuable tissue samples
Sectioning
  • Precision engineering for solid connection to microtomes
Archiving
  • Cassette surface texture optimizes scannability for archiving
Specifications
Specifications
Slots
Loose Packed
Separate Lid, Back Hinge
Green
Yes
1000/Cs.
No
Slotted Cassette