missing translation for 'onlineSavingsMsg'
Learn More

Epredia™ TissueLoc™ Cassette IV Slotted Cassettes

For routine tissue samples, cassettes have a back-edge lid hinge with loose-packed lids that are available in 12 different color varieties

140.00 €

Specifications

Closure Included Yes
Packaging Type Loose Packed
Quantity 1000/Cs.
Pore Style Slots
Closure Type Separate Lid, Back Hinge
View More Specs

Products 11
Product Code Brand Color Price Quantity & Availability  
Product Code Brand Color Price Quantity & Availability  
16304176
View Documents
Epredia™
B851954BE
Beige
140.00 €
Pack of 1000
Please to purchase this item. Need a web account? Register with us today!
16314176
View Documents
Epredia™
B851954BL
Blue
140.00 €
Pack of 1000
Please to purchase this item. Need a web account? Register with us today!
16324176
View Documents
Epredia™
B851954GN
Green
140.00 €
Pack of 1000
Please to purchase this item. Need a web account? Register with us today!
16344176
View Documents
Epredia™
B851954LA
Lavender
140.00 €
Pack of 1000
Please to purchase this item. Need a web account? Register with us today!
16354176
View Documents
Epredia™
B851954LG
Light Green
140.00 €
Pack of 1000
Please to purchase this item. Need a web account? Register with us today!
16364176
View Documents
Epredia™
B851954LI
Lilac
140.00 €
Pack of 1000
Please to purchase this item. Need a web account? Register with us today!
16374176
View Documents
Epredia™
B851954OR
Orange
140.00 €
Pack of 1000
Please to purchase this item. Need a web account? Register with us today!
16384176
View Documents
Epredia™
B851954PK
Pink
140.00 €
Pack of 1000
Please to purchase this item. Need a web account? Register with us today!
16394176
View Documents
Epredia™
B851954TN
Tan
140.00 €
Pack of 1000
Please to purchase this item. Need a web account? Register with us today!
16304186
View Documents
Epredia™
B851954WH
White
140.00 €
Pack of 1000
Please to purchase this item. Need a web account? Register with us today!
16314186
View Documents
Epredia™
B851954YW
Yellow
140.00 €
Pack of 1000
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Printing

  • Surface finish and reinforced print faces are engineered to optimize print quality
  • Tube design is precisely engineered to reduce jamming and prevent downtime

Grossing, Processing and Embedding

  • Cassette design and flow characteristics tested to ensure efficient reagent exchange
  • Opening and closing forces adjusted for ease of use, while safeguarding valuable tissue samples

Sectioning

  • Precision engineering for solid connection to microtomes

Archiving

  • Cassette surface texture optimizes scannability for archiving
Specifications

Specifications

Yes
1000/Cs.
Separate Lid, Back Hinge
Slotted Cassette
Loose Packed
Slots
No
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.