missing translation for 'onlineSavingsMsg'
Learn More

Hanna Instruments™ Waterproof Portable pH Meter

Features a large, dual level LCD display for easy viewing of the pH, temperature, and status indicators. Hanna Instruments™ Waterproof Portable pH Meter offers a simplified two-button operation. Compact, waterproof, and portable.

Brand:  Hanna Instruments™ HI991001

Product Code. 10088591

  • 864.24 € / Each
Estimated Shipment: 02-05-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

Compact, waterproof, portable pH/temperature meter that is supplied complete including a rugged carrying case and a unique titanium body pH electrode that has a built-in temperature sensor.
  • Large, dual level LCD display for easy viewing of the pH, temperature, and status indicators
  • Simplified two-button operation
  • Exceptionally long battery life of 1200 hours of continuous use
Specifications

Specifications

±0.5°C (up to 60°C); ±1.0°C (outside), ±1°F (up to 140°F);
Automatic one or two point pH calibration with auto buffer recognition
0.01
-5°C to +105°C
0.1°C, 0.1°F
152 x 58 x 30 mm
Yes
0.01pH
Automatic from -5 to 105ºC°C
Pre-amplified pH/ORP probe with internal
1.5 V
±0.02
-2 to 16
4.01, 7.01, 10.01 or 4.01, 6.86, 9.18
23°F to 221°F
LCD
HI991001
Cable
-2.0 to 16.0 pH, 1.5 V, -5.5°C to +105.0°C/23.0°F to 221.0°F Temperature
1.5V
1200 hr.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

One moment while we fetch your results.
Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Hanna Instruments™ Waterproof Portable pH Meter > Weight: 205g

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.