Learn More
BRAND™ Optical Glass Cavity Slides
With rectangular ground edges.
Brand:  BRAND™ 475565
 View more versions of this product
Product Code. 10151391
  • 337.00 € / Pack of 50
In Stock
to see stock.
Explore more special offers
Add to basket
Description
Description
  • White (clear) soda-lime glass, hydrolytic class 3
  • Single concavity
  • Provided in packs of 50
TRUSTED_SUSTAINABILITY
Specifications
Specifications
50
Soda Lime Glass
Optical Glass Slide
Clear White
Ground
1.2 to 1.5 mm
3
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
BRAND™ Optical Glass Cavity Slides > Well Count: 3