missing translation for 'onlineSavingsMsg'
Learn More

Titanium carbide, 99.5% (metals basis), Thermo Scientific Chemicals

69.65 € - 652.25 €

Chemical Identifiers

CAS 12070-08-5
Molecular Formula CTi
Molecular Weight (g/mol) 59.88
MDL Number MFCD00011268
InChI Key YXIVWSJCLXKLJL-UHFFFAOYSA-N
IUPAC Name methanidylidynetitaniumylium
SMILES [C-]#[Ti+]
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11359307
View Documents
Thermo Scientific Alfa Aesar
040178.18
50 g
69.65 €
50g
Please to purchase this item. Need a web account? Register with us today!
11339307
View Documents
Thermo Scientific Alfa Aesar
040178.30
250 g
191.96 €
250g
Please to purchase this item. Need a web account? Register with us today!
11349307
View Documents
Thermo Scientific Alfa Aesar
040178.A1
1 kg
652.25 €
1kg
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Used in preparation of cermets and abrasion-resistant surface coating on metal parts.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

12070-08-5
59.88
YXIVWSJCLXKLJL-UHFFFAOYSA-N
[C-]#[Ti+]
CTi
MFCD00011268
methanidylidynetitaniumylium

Specifications

4820°C
12070-08-5
99.5% (metals basis)
MFCD00011268
YXIVWSJCLXKLJL-UHFFFAOYSA-N
methanidylidynetitaniumylium
59.91
Titanium carbide
∽3140°C
Powder
CTi
Insoluble in water.
[C-]#[Ti+]
59.88
4.93 g/mL
Videos
Safety and Handling

Safety and Handling

P210-P240-P241-P280-P370+P378q

H228

missing translation for 'dotInformation' : Hazard Class: 4.1; Packaging Group: III

missing translation for 'einecsNumber' : 235-120-4

missing translation for 'rtecsNumber' : XR1903500

missing translation for 'tsca' : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.