Learn More
Titanium Granules, -15 mesh, 99.9% (Metals basis excluding Na and K), Thermo Scientific™
37.00 € - 448.82 €
Chemical Identifiers
CAS | 7440-32-6 |
---|---|
Molecular Formula | Ti |
Molecular Weight (g/mol) | 47.87 |
MDL Number | MFCD00011264,MFCD00151301 |
InChI Key | RTAQQCXQSZGOHL-UHFFFAOYSA-N |
Synonym | oremet, alloy, cp, vt1, c.p., contimet 30, jistp28, titanium-125, ii hydride, titan vt 1-1 |
PubChem CID | 23963 |
ChEBI | CHEBI:33341 |
IUPAC Name | titanium |
SMILES | [Ti] |
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Chemical Identifiers
7440-32-6 | |
47.87 | |
RTAQQCXQSZGOHL-UHFFFAOYSA-N | |
23963 | |
titanium |
Ti | |
MFCD00011264,MFCD00151301 | |
oremet, alloy, cp, vt1, c.p., contimet 30, jistp28, titanium-125, ii hydride, titan vt 1-1 | |
CHEBI:33341 | |
[Ti] |
Specifications
Gray | |
Dark Gray granules | |
Ti | |
oremet, alloy, cp, vt1, c.p., contimet 30, jistp28, titanium-125, ii hydride, titan vt 1-1 | |
[Ti] | |
47.87 | |
CHEBI:33341 | |
Odorless | |
Titanium |
7440-32-6 | |
99.9% (Metals basis excluding Na and K) | |
MFCD00011264,MFCD00151301 | |
RTAQQCXQSZGOHL-UHFFFAOYSA-N | |
titanium | |
23963 | |
-15 Mesh | |
Granular |
Safety and Handling
missing translation for 'einecsNumber' : 231-142-3
missing translation for 'rtecsNumber' : XR1700000
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
Your input is important to us. Please complete this form to provide feedback related to the content on this product.